Methyl-3-(hydroxymethyl)benzoat 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-(hydroxymethyl)benzoate

OR931154-1G
APOSOR931154-1GEA 68.3 EUR
APOSOR931154-1G
Methyl-3-(hydroxymethyl)benzoat 95%
Methyl-3-(hydroxymethyl)benzoat
Formel: C₉H₁₀O₃
Molecular Weight: 166,1739 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06202752
CAS-Nummer: 67853-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR