Bis(trimethylsilyl)carbodiimid 95%

Lieferant: Apollo Scientific

OR931437-25G OR931437-5G
APOSOR931437-25GEA 828 EUR
APOSOR931437-25G APOSOR931437-5G
Bis(trimethylsilyl)carbodiimid 95%
Bis(trimethylsilyl)carbodiimid
Formel: C₇H₁₈N₂Si₂
Molecular Weight: 186,4 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051538
CAS-Nummer: 1000-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR