2-Acetamidopyridine 95%

Lieferant: Apollo Scientific

OR931544-5G
APOSOR931544-5GEA 111 EUR
APOSOR931544-5G
2-Acetamidopyridine 95%
2-Acetamidopyridine
Formel: C₇H₈N₂O
Molecular Weight: 136.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00023423
CAS-Nummer: 5231-96-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR