2-Chlor-4,8-dimethylchinolin 98%

Lieferant: Apollo Scientific

OR931562-1G OR931562-5G
APOSOR931562-1GEA 36.4 EUR
APOSOR931562-1G APOSOR931562-5G
2-Chlor-4,8-dimethylchinolin 98%
2-Chloro-4,8-dimethylquinoline
Formel: C₁₁H₁₀ClN
Molecular Weight: 191,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00760267
CAS-Nummer: 3913-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR