4'-Amino-3'-cyanacetophenon 95%

Lieferant: Apollo Scientific

Synonyme: 5-Acetyl-2-aminobenzonitrile

OR931672-5G
APOSOR931672-5GEA 253 EUR
APOSOR931672-5G
4'-Amino-3'-cyanacetophenon 95%
4'-Amino-3'-cyanacetophenon
Formel: C₉H₈N₂O
Molecular Weight: 160,1726 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18379155
CAS-Nummer: 33720-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR