2-Methylisocrotonsäure-Isobutylester 95%

Lieferant: Apollo Scientific

Synonyme: (Z)-2-Methyl-2-butenoic Acid Isobutyl Ester, 2-Methylisocrotonic Acid Isobutyl Ester, (Z)-Isobutyl 2-methylbut-2-enoate, Angelic Acid Isobutyl Ester, Isobutyl (Z)-2-Methyl-2-butenoate

OR931863-25G OR931863-5G
APOSOR931863-25GEA 418 EUR
APOSOR931863-25G APOSOR931863-5G
2-Methylisocrotonsäure-Isobutylester 95%
Isobutyl angelate
Formel: C₉H₁₆O₂
Molecular Weight: 156,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00063646
CAS-Nummer: 7779-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR