(2,6-Dimethylphenoxy)essigsäure 98%

Lieferant: Apollo Scientific

Synonyme: 2-(2,6-Dimethylphenoxy)acetic acid

OR931889-100G OR931889-25G
APOSOR931889-100GEA 198 EUR
APOSOR931889-100G APOSOR931889-25G
(2,6-Dimethylphenoxy)essigsäure 98%
2,6-Dimethylphenoxyacetic acid
Formel: C₁₀H₁₂O₃
Molecular Weight: 180.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00156912
CAS-Nummer: 13335-71-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR