N-[(3,5-Dimethylisoxazol-4-yl)methyl]-N-methylamine 95%
Lieferant: Apollo Scientific
Synonyme:
N-[(3,5-Dimethylisoxazol-4-yl)methyl]-N-methylamine hydrochloride
OR932076-1G
OR932076-250MG
APOSOR932076-1GEA
574
EUR
APOSOR932076-1G
APOSOR932076-250MG
N-[(3,5-Dimethylisoxazol-4-yl)methyl]-N-methylamine 95%
N-[(3,5-Dimethylisoxazol-4-yl)methyl]-N-methylamine
Formel:
C₇H₁₂N₂O Molecular Weight: 140.19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08701362 CAS-Nummer: 893641-32-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...