1-Acetoxy-3-chloroacetone 96%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Chloro-2-oxopropyl acetate, 1-Acetoxy-3-acetenyl Chloride, 1-Acetoxy-3-chloro-2-propanone

OR932209-25G
APOSOR932209-25GEA 188 EUR
APOSOR932209-25G
1-Acetoxy-3-chloroacetone 96%
1-Acetoxy-3-chloroacetone
Formel: C₅H₇ClO₃
Molecular Weight: 150,56 g/mol
Dichte: 1,28 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00191365
CAS-Nummer: 40235-68-5
UN: 1760
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR