1-Bromo-3,3-dimethylbutane 98%
Lieferant: Apollo Scientific
OR932225-100G
APOSOR932225-100GEA
120
EUR
APOSOR932225-100G
1-Bromo-3,3-dimethylbutane 98%
1-Bromo-3,3-dimethylbutane
Formel:
C₆H₁₃Br Molecular Weight: 165,07 g/mol Siedepunkt: 134…140 °C Dichte: 1,1556 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00060844 CAS-Nummer: 1647-23-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...