2,1-Benzisoxazol 98%

Lieferant: Apollo Scientific

Synonyme: Anthranil, Benzo[c]isoxazole, 2,1-Benzisoxazole

OR932514-5G
APOSOR932514-5GEA 111 EUR
APOSOR932514-5G
2,1-Benzisoxazol 98%
2,1-Benzisoxazole
Formel: C₇H₅NO
Molecular Weight: 119,1207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005806
CAS-Nummer: 271-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR