3-Chloro-N-methylpyridin-2-amine 95%

Lieferant: Apollo Scientific

OR932541-1G OR932541-5G
APOSOR932541-1GEA 328 EUR
APOSOR932541-1G APOSOR932541-5G
3-Chloro-N-methylpyridin-2-amine 95%
3-Chloro-N-methylpyridin-2-amine
Formel: C₆H₇ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11122502
CAS-Nummer: 468718-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR