Methyl-2-(brommethyl)-6-nitrobenzoat 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Methyl 2-(bromomethyl)-6-nitrobenzoate

OR932561-250MG OR932561-1G
APOSOR932561-250MGEA 171 EUR
APOSOR932561-250MG APOSOR932561-1G
Methyl-2-(brommethyl)-6-nitrobenzoat 98%
Methyl-2-(brommethyl)-6-nitrobenzoat
Formel: C₉H₈BrNO₄
Molecular Weight: 274,0700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114314
CAS-Nummer: 61940-21-4
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR