2-Octen-4-on 95%
Lieferant: Apollo Scientific
OR932607-1G
OR932607-5G
APOSOR932607-1GEA
203
EUR
APOSOR932607-1G
APOSOR932607-5G
2-Octen-4-on 95%
2-Octen-4-one
Formel:
C₈H₁₄O Molecular Weight: 126,20 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061023 CAS-Nummer: 4643-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...