Methyl-4-(4-aminophenyl)butanoate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 4-(4-aminophenyl)butanoate
OR932646-1G
OR932646-5G
APOSOR932646-1GEA
94.4
EUR
APOSOR932646-1G
APOSOR932646-5G
Methyl-4-(4-aminophenyl)butanoate 95%
Methyl-4-(4-aminophenyl)butanoate
Formel:
C₁₁H₁₅NO₂ Molecular Weight: 193,2400 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07366753 CAS-Nummer: 20637-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...