4-Benzoylbutyric acid 95%

Lieferant: Apollo Scientific

OR932647-100G OR932647-25G OR932647-5G
APOSOR932647-100GEA 0 EUR
APOSOR932647-100G APOSOR932647-25G APOSOR932647-5G
4-Benzoylbutyric acid 95%
4-Benzoylbutyric acid
Formel: C₁₁H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00004411
CAS-Nummer: 1501-05-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR