2-Buten-1-ylsuccinic anhydride 95%

Lieferant: Apollo Scientific

Synonyme: 4-Hexene-1,2-dicarboxylic Anhydride

OR932680-1G OR932680-25G OR932680-5G
APOSOR932680-1GEA 145 EUR
APOSOR932680-1G APOSOR932680-25G APOSOR932680-5G
2-Buten-1-ylsuccinic anhydride 95%
2-Buten-1-ylsuccinic anhydride
Formel: C₈H₁₀O₃
Molecular Weight: 154,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03423563
CAS-Nummer: 7538-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR