4-(2-Methoxyethyl)phenol 97%
Lieferant: Apollo Scientific
Synonyme:
p-(2-Methoxyethyl) phenol
OR932723-500G
APOSOR932723-500GEA
183
EUR
APOSOR932723-500G
4-(2-Methoxyethyl)phenol 97%
4-(2-Methoxyethyl)phenol
Formel:
C₉H₁₂O₂ Molecular Weight: 152,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017537 CAS-Nummer: 56718-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...