4-Propoxyphenol 95%

Lieferant: Apollo Scientific

Synonyme: Hydrochinonmonopropylether

OR932741-100G OR932741-25G
APOSOR932741-100GEA 426 EUR
APOSOR932741-100G APOSOR932741-25G
4-Propoxyphenol 95%
4-Propoxyphenol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002335
CAS-Nummer: 18979-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR