Genipin 98%
Lieferant: Apollo Scientific
Synonyme:
Methyl (1S,2R,6S)-2-Hydroxy-9-(hydroxymethyl)-3-oxabicyclo[4.3.0]nona-4,8-diene-5-carboxylate, (1R,4aS,7aS)-Methyl 1-hydroxy-7-(hydroxymethyl)-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-4-carboxylate
OR932863-1G
OR932863-5G
APOSOR932863-1GEA
23.2
EUR
APOSOR932863-1G
APOSOR932863-5G
Genipin 98%
Genipin
Formel:
C₁₁H₁₄O₅ Molecular Weight: 226,23 g/mol Schmelzpunkt: 120…121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00888600 CAS-Nummer: 6902-77-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...