4-(4-Bromophenyl)-1-tert-butoxycarbonylpiperidine-4-carboxylic acid 97%
Lieferant: Apollo Scientific
Synonyme:
1-Boc-4-(4-bromophenyl)-4-carboxypiperidine
OR932876-1G
OR932876-250MG
OR932876-500MG
APOSOR932876-1GEA
411
EUR
APOSOR932876-1G
APOSOR932876-250MG
APOSOR932876-500MG
4-(4-Bromophenyl)-1-tert-butoxycarbonylpiperidine-4-carboxylic acid 97%
4-(4-Bromophenyl)-1-tert-butoxycarbonylpiperidine-4-carboxylic acid
Formel:
C₁₇H₂₂BrNO₄ Molecular Weight: 384,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226814 CAS-Nummer: 1076197-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...