Methyl-1-indolcarboxylat 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 1H-indole-1-carboxylate
OR933004-1G
OR933004-5G
APOSOR933004-1GEA
190
EUR
APOSOR933004-1G
APOSOR933004-5G
Methyl-1-indolcarboxylat 95%
Methyl-1-indolcarboxylat
Formel:
C₁₀H₉NO₂ Molecular Weight: 175,19 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
39203-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...