4-Methoxy-2-methylphenol 96%

Lieferant: Apollo Scientific

Synonyme: 4-Hydroxy-3-methylanisol

OR933049-10G OR933049-5G OR933049-250MG
APOSOR933049-10GEA 368 EUR
APOSOR933049-10G APOSOR933049-5G APOSOR933049-250MG
4-Methoxy-2-methylphenol 96%
4-Methoxy-2-methylphenol
Formel: C₈H₁₀O₂
Molecular Weight: 138,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11055138
CAS-Nummer: 5307-05-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR