[4-(Dimethylamino)phenyl]acetic acid 95%

Lieferant: Apollo Scientific

Synonyme: 4-(Dimethylamino)phenylacetic acid

OR933243-1G OR933243-250MG OR933243-5G
APOSOR933243-1GEA 114 EUR
APOSOR933243-1G APOSOR933243-250MG APOSOR933243-5G
[4-(Dimethylamino)phenyl]acetic acid 95%
[4-(Dimethylamino)phenyl]acetic acid
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00075591
CAS-Nummer: 17078-28-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR