2-Chloro-3-nitropyridin-4-ol 95%

Lieferant: Apollo Scientific

OR933369-1G OR933369-250MG
APOSOR933369-1GEA 708 EUR
APOSOR933369-1G APOSOR933369-250MG
2-Chloro-3-nitropyridin-4-ol 95%
2-Chloro-3-nitropyridin-4-ol
Formel: C₅H₃ClN₂O₃
Molecular Weight: 174.5400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07778466
CAS-Nummer: 629655-23-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR