2',6'-Dimethylacetophenon 97%

Lieferant: Apollo Scientific

Synonyme: 1-(2,6-Dimethylphenyl)ethan-1-on

OR933402-5G
APOSOR933402-5GEA 82.8 EUR
APOSOR933402-5G
2',6'-Dimethylacetophenon 97%
2',6'-Dimethylacetophenon
Formel: C₁₀H₁₂O
Molecular Weight: 148,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01861394
CAS-Nummer: 2142-76-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR