2-(Dimethylaminomethyl)-3-hydroxypyridine 95%
Lieferant: Apollo Scientific
Synonyme:
2-((Dimethylamino)methyl)pyridin-3-ol
OR933407-25G
APOSOR933407-25GEA
197
EUR
APOSOR933407-25G
2-(Dimethylaminomethyl)-3-hydroxypyridine 95%
2-(Dimethylaminomethyl)-3-hydroxypyridine
Formel:
C₈H₁₂N₂O Molecular Weight: 152,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006347 CAS-Nummer: 2168-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...