5-(2-Thienyl)-1H-pyrazole
Lieferant: Apollo Scientific
Synonyme:
3-(Thiophen-2-yl)-1H-pyrazole
OR9336-1G
OR9336-250MG
APOSOR9336-1GEA
33.5
EUR
APOSOR9336-1G
APOSOR9336-250MG
5-(2-Thienyl)-1H-pyrazole
5-(2-Thienyl)-1H-pyrazole
Formel:
C₇H₆N₂S Molecular Weight: 150.2 g/mol Siedepunkt: 152 °C/1.6mmHg Schmelzpunkt: 97…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059740 CAS-Nummer: 19933-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...