3-(2,3-Dihydroindol-1-yl)propionic acid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Indolin-1-yl)propanoic acid

OR9339-1G OR9339-5G OR9339-25G
APOSOR9339-1GEA 136 EUR
APOSOR9339-1G APOSOR9339-5G APOSOR9339-25G
3-(2,3-Dihydroindol-1-yl)propionic acid 95+%
3-(2,3-Dihydroindol-1-yl)propionic acid
Formel: C₁₁H₁₃NO₂
Molecular Weight: 191,23 g/mol
Schmelzpunkt: 75…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00220603
CAS-Nummer: 99855-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR