2,4-Dimethyl-5-thiazolcarbonsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Dimethylthiazole-5-carboxylic acid

OR9354-1G OR9354-10G OR9354-5G
APOSOR9354-1GEA 82.9 EUR
APOSOR9354-1G APOSOR9354-10G APOSOR9354-5G
2,4-Dimethyl-5-thiazolcarbonsäure 98%
2,4-Dimethyl-5-thiazolecarboxylic acid
Formel: C₆H₇NO₂S
Molecular Weight: 157.1900 g/mol
Schmelzpunkt: 233…235 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00052943
CAS-Nummer: 53137-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR