3-(1-Adamantyl)-4-methoxybenzeneboronic acid 96%

Lieferant: Apollo Scientific

Synonyme: (3-(Adamantan-1-yl)-4-methoxyphenyl)boronic acid

OR9355-1G OR9355-250MG
APOSOR9355-1GEA 159 EUR
APOSOR9355-1G APOSOR9355-250MG
3-(1-Adamantyl)-4-methoxybenzeneboronic acid 96%
3-(1-Adamantyl)-4-methoxybenzeneboronic acid
Formel: C₁₇H₂₃BO₃
Molecular Weight: 286.18 g/mol
Schmelzpunkt: 239…242 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08056355
CAS-Nummer: 459423-32-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR