1-(2-Chlorethyl)pyrrol 95%
Lieferant: Apollo Scientific
Synonyme:
1-(2-Chloroethyl)-1H-pyrrole
OR936206-1G
OR936206-5G
APOSOR936206-1GEA
63.8
EUR
APOSOR936206-1G
APOSOR936206-5G
1-(2-Chlorethyl)pyrrol 95%
1-(2-Chloroethyl)pyrrole
Formel:
C₆H₈ClN Molecular Weight: 129,5874 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191330 CAS-Nummer: 77200-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...