(S)-(-)-1-Methyl-2-(1-piperidinomethyl)pyrrolidine 95%

Lieferant: Apollo Scientific

Synonyme: (S)-(-)-1-[(1-Methyl-2-pyrrolidinyl)methyl]piperidine

OR936287-1G
APOSOR936287-1GEA 85.7 EUR
APOSOR936287-1G
(S)-(-)-1-Methyl-2-(1-piperidinomethyl)pyrrolidine 95%
(S)-(-)-1-Methyl-2-(1-piperidinomethyl)pyrrolidine
Formel: C₁₁H₂₂N₂
Molecular Weight: 182,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077821
CAS-Nummer: 84466-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR