Copper(I)-2-thiophenecarboxylate 95%

Lieferant: Apollo Scientific

Synonyme: (2-Thiophenecarboxylato)copper(I), Copper(I) thiophene-2-carboxylate, 2-Thiophenecarboxylic Acid Copper(I) Salt, CuTC

OR936291-100G OR936291-25G
APOSOR936291-100GEA 161 EUR
APOSOR936291-100G APOSOR936291-25G
Copper(I)-2-thiophenecarboxylate 95%
Copper(I)-2-thiophenecarboxylate
Formel: C₅H₃CuO₂S
Molecular Weight: 190.68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02183524
CAS-Nummer: 68986-76-5
UN: 3077
ADR: 9+EHS III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR