(±)-3-Phenylbuttersäure 98%
Lieferant: Apollo Scientific
Synonyme:
(±)-3-Phenylbutansäure
OR936346-5G
APOSOR936346-5GEA
102
EUR
APOSOR936346-5G
(±)-3-Phenylbuttersäure 98%
(±)-3-Phenylbuttersäure
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002725 CAS-Nummer: 4593-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...