4-tert-Butoxybenzaldehyd 95%

Lieferant: Apollo Scientific

Synonyme: 4-(tert-Butoxy)benzaldehyde

OR936350-1G OR936350-25G OR936350-5G
APOSOR936350-1GEA 31.9 EUR
APOSOR936350-1G APOSOR936350-25G APOSOR936350-5G
4-tert-Butoxybenzaldehyd 95%
4-tert-Butoxybenzaldehyd
Formel: C₁₁H₁₄O₂
Molecular Weight: 178,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00040800
CAS-Nummer: 57699-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR