N-(3-Chlorpropyl)-4-boronobenzolsulfonamid 98%

Lieferant: Apollo Scientific

Synonyme: 4-[N-(3-Chloropropyl)sulphamoyl]benzeneboronic acid

OR9364-1G OR9364-250MG
APOSOR9364-1GEA 430 EUR
APOSOR9364-1G APOSOR9364-250MG
N-(3-Chlorpropyl)-4-boronobenzolsulfonamid 98%
N-(3-Chlorpropyl)-4-boronobenzolsulfonamid
Formel: C₉H₁₃BClNO₄S
Schmelzpunkt: 243…245 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235065
CAS-Nummer: 874219-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR