1,4-Bis(2-hydroxyethyl)benzene 95%
Lieferant: Apollo Scientific
Synonyme:
1,4-Benzenediethanol, 1,4-Phenylenediethanol
OR936439-1G
OR936439-5G
APOSOR936439-1GEA
140
EUR
APOSOR936439-1G
APOSOR936439-5G
1,4-Bis(2-hydroxyethyl)benzene 95%
1,4-Bis(2-hydroxyethyl)benzene
Formel:
C₁₀H₁₄O₂ Molecular Weight: 166.22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00666548 CAS-Nummer: 5140-03-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...