N,N-Dipropylanilin 95%

Lieferant: Apollo Scientific

Synonyme: N-Phenyldipropylamine

OR936458-25G OR936458-5G
APOSOR936458-25GEA 183 EUR
APOSOR936458-25G APOSOR936458-5G
N,N-Dipropylanilin 95%
N,N-Dipropylanilin
Formel: C₁₂H₁₉N
Molecular Weight: 177,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00035797
CAS-Nummer: 2217-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR