Ethyl-2-(2-amino-4-thiazolyl)-2-(hydroxyimino)acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Amino-4-thiazolyl)-2-(hydroxyimino)acetic Acid Ethyl Ester

OR936499-25G OR936499-5G
APOSOR936499-25GEA 139 EUR
APOSOR936499-25G APOSOR936499-5G
Ethyl-2-(2-amino-4-thiazolyl)-2-(hydroxyimino)acetate
Ethyl-2-(2-amino-4-thiazolyl)-2-(hydroxyimino)acetate
Formel: C₇H₉N₃O₃S
Molecular Weight: 215,23 g/mol
Schmelzpunkt: 195…197 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010415
CAS-Nummer: 60845-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR