N,N-Dimethyl-2-naphthalinamin 95%
Lieferant: Apollo Scientific
Synonyme:
N,N-Dimethylnaphthalen-2-amine, 2-Dimethylaminonaphthalene
OR936521-5G
APOSOR936521-5GEA
149
EUR
APOSOR936521-5G
N,N-Dimethyl-2-naphthalinamin 95%
N,N-Dimethyl-2-naphthalinamin
Formel:
C₁₂H₁₃N Molecular Weight: 171,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059550 CAS-Nummer: 2436-85-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...