3-Phenyl-2,4-pentandion 95%

Lieferant: Apollo Scientific

Synonyme: 3-Phenylacetylacetone, 3-Phenylpentane-2,4-dione

OR936526-1G OR936526-250MG
APOSOR936526-1GEA 95.7 EUR
APOSOR936526-1G APOSOR936526-250MG
3-Phenyl-2,4-pentandion 95%
3-Phenyl-2,4-pentandion
Formel: C₁₁H₁₂O₂
Molecular Weight: 176,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00048223
CAS-Nummer: 5910-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR