1-Isopropyl-3-pyrrolidinol 95%

Lieferant: Apollo Scientific

Synonyme: 1-Isopropyl-3-hydroxypyrrolidine

OR936536-10G OR936536-1G OR936536-5G
APOSOR936536-10GEA 899 EUR
APOSOR936536-10G APOSOR936536-1G APOSOR936536-5G
1-Isopropyl-3-pyrrolidinol 95%
1-Isopropyl-3-pyrrolidinol
Formel: C₇H₁₅NO
Molecular Weight: 129,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191525
CAS-Nummer: 42729-56-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR