2'-Acetoxyacetophenon 97%
Lieferant: Apollo Scientific
Synonyme:
o-Acetylphenylacetat, 2-Acetylphenylacetat, Essigsäure(2-acetylphenyl)ester
OR936566-100G
OR936566-25G
APOSOR936566-100GEA
256
EUR
APOSOR936566-100G
APOSOR936566-25G
2'-Acetoxyacetophenon 97%
2'-Acetoxyacetophenon
Formel:
C₁₀H₁₀O₃ Molecular Weight: 178,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017223 CAS-Nummer: 7250-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...