4-Butylbenzaldehyd 95%

Lieferant: Apollo Scientific

OR936585-25G OR936585-5G
APOSOR936585-25GEA 843 EUR
APOSOR936585-25G APOSOR936585-5G
4-Butylbenzaldehyd 95%
4-Butylbenzaldehyd
Formel: C₁₁H₁₄O
Molecular Weight: 162,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00040751
CAS-Nummer: 1200-14-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR