Fumaraldehyd Mono(Dimethylacetal) 95%

Lieferant: Apollo Scientific

Synonyme: 4,4-Dimethoxycrotonaldehyde, 4,4-Dimethoxy-2-butenal

OR936614-1G OR936614-250MG
APOSOR936614-1GEA 654 EUR
APOSOR936614-1G APOSOR936614-250MG
Fumaraldehyd Mono(Dimethylacetal) 95%
Fumaraldehyde mono(dimethyl acetal)
Formel: C₆H₁₀O₃
Molecular Weight: 130,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00191439
CAS-Nummer: 4093-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR