2-Chlor-N,N-diethylanilin 95%

Lieferant: Apollo Scientific

OR936656-10G OR936656-1G OR936656-5G
APOSOR936656-10GEA 790 EUR
APOSOR936656-10G APOSOR936656-1G APOSOR936656-5G
2-Chlor-N,N-diethylanilin 95%
2-Chloro-N,N-diethylaniline
Formel: C₁₀H₁₄ClN
Molecular Weight: 183,68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059477
CAS-Nummer: 19372-80-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR