(S)-(-)-β-Citronellol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-(-)-Citronellol

OR936732-25G OR936732-5G OR936732-100G
APOSOR936732-25GEA 78.3 EUR
APOSOR936732-25G APOSOR936732-5G APOSOR936732-100G
(S)-(-)-β-Citronellol 95%
(S)-(-)-β-Citronellol
Formel: C₁₀H₂₀O
Molecular Weight: 156.27 g/mol
Siedepunkt: 225…226 °C
Dichte: 0.856 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00063214
CAS-Nummer: 7540-51-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR