Ethylmethylsulfon 95%
Lieferant: Apollo Scientific
Synonyme:
Ethylmethylsulphone, Methylsulphonylethane
OR936735-1G
OR936735-25G
OR936735-5G
APOSOR936735-1GEA
21.9
EUR
APOSOR936735-1G
APOSOR936735-25G
APOSOR936735-5G
Ethylmethylsulfon 95%
Ethyl methyl sulphone
Formel:
C₃H₈O₂S Molecular Weight: 108,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015543 CAS-Nummer: 594-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...