4-Butoxybenzylalkohol 95%

Lieferant: Apollo Scientific

Synonyme: (4-Butoxyphenyl)methanol

OR936762-25G
APOSOR936762-25GEA 148 EUR
APOSOR936762-25G
4-Butoxybenzylalkohol 95%
4-Butoxybenzylalkohol
Formel: C₁₁H₁₆O₂
Molecular Weight: 180,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004657
CAS-Nummer: 6214-45-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR